Learn More
Microsolv Technology Corporation Freezer Storage Caps For Vials
Catalog No. NC1865840
Encompass_Preferred
Caps, Screw Top, Solid, with an ultra-pure, Silicone Rubber / PTFE liner, in a closed Black Cap. For use with 9-425mm thread,12x32mm autosampler vials. AQR Brand 100/PK.
Catalog No. NC1865840 Supplier Microsolv Technology Corporation Supplier No. 9509S10CKSR
Caps, Screw Top, Solid, with an ultra-pure, Silicone Rubber / PTFE liner, in a closed Black Cap. For use with 9-425mm thread,12x32mm autosampler vials. AQR Brand 100/PK.