Learn More
PHCbi VIP ECO SMART -86°C Freezer
Catalog No. NC3093449
Encompass_Preferred
NEW! Independently tested and documented, the ENERGY STAR Certified VIP ECO SMART delivers the lowest daily energy usage (5.40 KWh per day) of any ultra-low temperature freezer in its class. The dual voltage MDF-DU703VHA-PA is purpose-designed for the temperature uniformity and stability required for demanding life sciences applications. It runs on either 115V or 220V power using the corresponding, removable power cords that come standard with every freezer. Variable speed inverter-driven compressors with proprietary SMART control algorithms allow for superior uniformity by running compressors at a slower speed under steady-state conditions.
Catalog No. NC3093449 Supplier PHCbi Supplier No. MDFDU703VHAPA
Add to Cart
Add to Cart
Request a Quote
NEW! Independently tested and documented, the ENERGY STAR Certified VIP ECO SMART delivers the lowest daily energy usage (5.40 KWh per day) of any ultra-low temperature freezer in its class. The dual voltage MDF-DU703VHA-PA is purpose-designed for the temperature uniformity and stability required for demanding life sciences applications. It runs on either 115V or 220V power using the corresponding, removable power cords that come standard with every freezer. Variable speed inverter-driven compressors with proprietary SMART control algorithms allow for superior uniformity by running compressors at a slower speed under steady-state conditions.