Learn More
Shockwatch Div Mri FreezeSafe 0°C
SDP
Catalog No. NC2598344
Encompass_Preferred
FreezeSafe temperature indicator monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably low temperature levels, allowing you to determine if product quality has been compromised. Temperature threshold: 0C/32F. Indicator turns from clear to magenta withing 60 minutes. Accuracy +/-1C. Shelf life 12 months. Single-use.
Catalog No. NC2598344 Supplier Shockwatch Div Mri Supplier No. E01FRE000C02P1
FreezeSafe temperature indicator monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably low temperature levels, allowing you to determine if product quality has been compromised. Temperature threshold: 0C/32F. Indicator turns from clear to magenta withing 60 minutes. Accuracy +/-1C. Shelf life 12 months. Single-use.