Learn More
ATHENA ES FREEZING MEDIA PAIR 50ML
Catalog No. NC0608215
Encompass_Preferred
Athena’s Freezing Media PairTM are a dual-cryopreservation system used to safely store suspensions of viable cells detached from monolayer cultures using the “cold trypsin” method with Athena’s cell dissociation media, PETTM. The pair of solutions thoroughly preserve cells which provides superior cell viability upon reconstitution. This Freezing Media PairTM has been used extensively to cryopreserve a variety of human cell lines grown in serum-free media, including breast and prostate cancer cell lines.
Catalog No. NC0608215 Supplier ATHENA ES Supplier No. 0406
Athena’s Freezing Media PairTM are a dual-cryopreservation system used to safely store suspensions of viable cells detached from monolayer cultures using the “cold trypsin” method with Athena’s cell dissociation media, PETTM. The pair of solutions thoroughly preserve cells which provides superior cell viability upon reconstitution. This Freezing Media PairTM has been used extensively to cryopreserve a variety of human cell lines grown in serum-free media, including breast and prostate cancer cell lines.