Learn More
NEXPRING HEALTH Freezing Medium - TYB with Glycerol & Gentamicin, 20 x 5 mL
Catalog No. NC9632816
Encompass
Freezing Medium TYB contains TEST-yolk buffer (TYB) to recover more motile sperm post-thaw than glycerol-only media (Hallak 2000, Nallella 2004). TEST-yolk buffer is a semen extender and cryobuffer that can be paired with glycerol to achieve high sperm motility and viability. The egg yolk is from USDA certified virus-free flocks and washed from the sperm after cryopreservation. Freezing Medium TYB is also the only CE-marked and FDA-approved sperm freezing media containing TYB and glycerol. As one of the most referenced sperm cryopreservation media worldwide, Freezing Medium TYB has established itself as the standard for performance.
Catalog No. NC9632816 Supplier NEXPRING HEALTH Supplier No. 9012820X5ML
Freezing Medium TYB contains TEST-yolk buffer (TYB) to recover more motile sperm post-thaw than glycerol-only media (Hallak 2000, Nallella 2004). TEST-yolk buffer is a semen extender and cryobuffer that can be paired with glycerol to achieve high sperm motility and viability. The egg yolk is from USDA certified virus-free flocks and washed from the sperm after cryopreservation. Freezing Medium TYB is also the only CE-marked and FDA-approved sperm freezing media containing TYB and glycerol. As one of the most referenced sperm cryopreservation media worldwide, Freezing Medium TYB has established itself as the standard for performance.