Learn More
Sigma Aldrich Fine Chemicals Biosciences Freezing vials, natural, 500/pk
Supplier:  Sigma Aldrich Fine Chemicals Biosciences V4381500EA
Encompass
Manufactured from an exceptionally clear, virgin polypropylene with a medical-grade silicone O-ring seal. These vials are designed for safe storage at temperatures as low as -196°C but should be used only in the gas phase of liquid nitrogen.
Catalog No. NC1187537