Learn More
Sigma Aldrich Fine Chemicals Biosciences Freezing vials, natural, 500/pk
Catalog No. NC1766010
Encompass
Manufactured from an exceptionally clear, virgin polypropylene with a medical-grade silicone O-ring seal. These vials are designed for safe storage at temperatures as low as -196°C but should be used only in the gas phase of liquid nitrogen. Capacity 500 uL, skirted bottom, sterile.
Catalog No. NC1766010 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. V3381500EA
Manufactured from an exceptionally clear, virgin polypropylene with a medical-grade silicone O-ring seal. These vials are designed for safe storage at temperatures as low as -196°C but should be used only in the gas phase of liquid nitrogen. Capacity 500 uL, skirted bottom, sterile.