Learn More
Electron Microscopy Sciences Mr. Frosty Cryo 1° C Freezing Container, 4.5-5.0 ml
SDP
Catalog No. 501928545
Encompass_Preferred
Mr. Frosty Cryo 1 degree C freezing containers are inexpensive, reusable, and convenient devices for freezing biological samples. The simple-to-use system is designed to achieve a rate of cooling very close to -1 degree C/minute, the optimal rate for cell preservation. Isopropyl alcohol is necessary to achieve the recommended rate of cooling. Holds 12 to 18 tubes.
Volume: 4.5-5.0 ml
Catalog No. 50-192-8545 Supplier Electron Microscopy Sciences Supplier No. 7259603
Mr. Frosty Cryo 1 degree C freezing containers are inexpensive, reusable, and convenient devices for freezing biological samples. The simple-to-use system is designed to achieve a rate of cooling very close to -1 degree C/minute, the optimal rate for cell preservation. Isopropyl alcohol is necessary to achieve the recommended rate of cooling. Holds 12 to 18 tubes.
Volume: 4.5-5.0 ml