Learn More
Glen Mills Inc French Press G-M
SDP
Catalog No. nc0355680
Encompass
Matches the performance of traditional FRENCH PRESSES. Old style Standard PRESSURE CELLS (40 kpsi; 35ml) can be used. Sold separately. Highest pressure hydraulic press commercially available to disrupt the toughest cells. Internal pressures generated within the Standard PRESSURE CELL can attain 40,000 psi. Sample volumes to 35ml with Standard PRESSURE CELL; minimal sample loss. Highly Polished Mirror Finish Stainless Steel Housing: 150 grit electro polished for sanitary applications and for ease of cleaning. Large easy to read 6” Hydraulic Pressure Gauge, mounted prominently on front. Pressure limits clearly marked on Pressure Gauge. Pressure Regulator Valve offers good control, easy to operate. Ratio Selector Switch is non-mechanical, twin solenoids for long life. Forced air intercooler for hydraulic fluid to maintain proper fluid viscosity. Tables for Hydraulic Pressure Gauge reading conversion to Inner PRESSURE CELL pressure conveniently located on front panel.
Catalog No. NC0355680 Supplier Glen Mills Inc Supplier No. 5500000011
Add to Cart
Add to Cart
Request a Quote
Matches the performance of traditional FRENCH PRESSES. Old style Standard PRESSURE CELLS (40 kpsi; 35ml) can be used. Sold separately. Highest pressure hydraulic press commercially available to disrupt the toughest cells. Internal pressures generated within the Standard PRESSURE CELL can attain 40,000 psi. Sample volumes to 35ml with Standard PRESSURE CELL; minimal sample loss. Highly Polished Mirror Finish Stainless Steel Housing: 150 grit electro polished for sanitary applications and for ease of cleaning. Large easy to read 6” Hydraulic Pressure Gauge, mounted prominently on front. Pressure limits clearly marked on Pressure Gauge. Pressure Regulator Valve offers good control, easy to operate. Ratio Selector Switch is non-mechanical, twin solenoids for long life. Forced air intercooler for hydraulic fluid to maintain proper fluid viscosity. Tables for Hydraulic Pressure Gauge reading conversion to Inner PRESSURE CELL pressure conveniently located on front panel.