Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Freon™ Trichlorofluoromethane, MilliporeSigma™ Supelco™

Catalog No. 111018702 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8702 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8702 Supplier MilliporeSigma Supelco Supplier No. 48541
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line Freon
CAS 75-69-4
Vapor Pressure 39.17 psi (55°C)
Vapor Density 5.04 (vs. air)
Density 1.494 g/mL (at 25°C)
Physical Form Neat
Linear Formula Cl3 CF
MDL Number MFCD00000784
Melting Point -111°C to -110°C
Boiling Point 23.7°C
Synonym CFC-11; Fluorotrichloromethane; Freon(R) 11
Type Food and Cosmetic Component Standard
Quantity 5000 mg
Formula Weight 137.37
Grade Analytical Standard
Show More Show Less

Freon is a registered trademark of E. I. du Pont de Nemours and Company

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.