Learn More
Aviva Systems Biology Corp FREQ Antibody - middle region (ARP34711_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34711_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FREQ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FREQ. Alternate names: DKFZp761L1223, FLUP, NCS-1, NCS1, FREQApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-55527