Learn More
Electron Microscopy Sciences Forensic Certified SEM Mount 6 mm pin
SDP
Supplier:  Electron Microscopy Sciences 7647554
Encompass_Preferred
This mount is made from a special certified Aluminum alloy which does not contain any of the objectionable elements that may interfere with samples collected for GSR . These mounts are 12.7 mm x 11 mm.
Catalog No. 50-192-7155