Learn More
Medchemexpress LLC HY-15652A 5mg Medchemexpress, Freselestat (quarterhydrate) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15652A5MG
Encompass_Preferred
Medchemexpress, HY-15652A 5mg Freselestat (quarterhydrate) CAS:0 Freselestat quarterhydrate (ONO-6818 quarterhydrate) is a potent and orally active neutrophil elastase inhibitor with a Ki of 12.2 nM. Freselestat quarterhydrate is >100-fold less-active against other proteases such as trypsin, protein-ase 3, pancreatic elastase, plasmin, thrombin, collagenase, cathepsin G, and murine macrophage elastase. Freselestat quarterhydrate has a potent anti-inflammatory activity[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0661