Learn More
Marketlab, Inc. Fresh-Mix Bleach Dilution System
Description
- Keeps water and bleach in separate cartridges until the time of use
- Each bleach cartridge contains 5.25% sodium hypochlorite
- Highly effective against a broad spectrum of pathogens, including C. diff spores
- Prevents bleach from degrading
- Registered with the EPA
Specifications
Specifications
| Product Type | Trigger Spray Bottle |
| Color-coded | No |
| Form | Spray |
| Scent | Unscented |
| Sterility | Non-sterile |