Learn More
Enterprise Technology Solutions Fresh Products Liquid/Spill Absorber, Shake Can, 12oz, Lemon Scent
SDP
Catalog No. 5000829102
Encompass_Preferred
Super-Sorb's highly absorbent, cellulose-based formula contains spills quickly. It locks odors away and turns liquids into a semi-solid gel for fast, easy cleanup. Super-Sorb Granular Absorbent absorbs 60 times its weight almost immediately and up to 400 times its weight after only a few minutes. It works great on vomit, blood, urine and other smelly messes. Biodegradable gel with a fresh lemon scent is an environmentally friendly solution.
Catalog No. 50-008-29102 Supplier Enterprise Technology Solutions Supplier No. FRSSS24LE
Super-Sorb's highly absorbent, cellulose-based formula contains spills quickly. It locks odors away and turns liquids into a semi-solid gel for fast, easy cleanup. Super-Sorb Granular Absorbent absorbs 60 times its weight almost immediately and up to 400 times its weight after only a few minutes. It works great on vomit, blood, urine and other smelly messes. Biodegradable gel with a fresh lemon scent is an environmentally friendly solution.