Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Fresh Water Test Kit
SDPSCI_ED

Catalog No. S45078 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
Fresh Water Test Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S45078 Fresh Water Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S45078 Supplier LaMotte™ Supplier No. 590202
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Six individual cases that house components needed for various chemical tests
  • Sampling bottles
  • Analysis report forms
  • 50 tests per factor, except nitrate (40) and pH (100)
  • Three handbooks now available as a download: Study of Water Quality, Limnology, and Investigating Water Problems

Teaches sampling techniques, careful measurement and data recording.

  • Combines limnology with a water testing kit
  • Tests for dissolved oxygen, carbon dioxide, nitrates, phosphates, silica, pH, calcium, magnesium and total hardness
  • Soft-side tote is made of durable fabric and a solid plastic bottom ready for field work
  • Handy side pockets hold thermometers, pens, writing tablets and other tools
  • Three supplemental handbooks are now available as a download: Study of Water Quality, Limnology, and Investigating Water Problems

Octa-Slide 2 Comparator Tests

  • Nitrate Nitrogen, 0.2-1.0 ppm (40 tests), 3119-01
  • Phosphate, 0.2-1.0 ppm (50 tests), 3119-01
  • pH, 3.0-10.0 pH (50 tests), 5858-01
  • Silica, 0.5-10 ppm (50 tests), 4463-01

Direct Reading Titrator Tests

  • Carbon Dioxide, 0-50 ppm (50 tests), 7297-DR-01
  • Dissolved Oxygen, 0-10 ppm (50 tests), 5860-01
  • Hardness, 0-200 ppm (50 tests), 4824-DR-LT-01

Order Info

Grades 5+.

Specifications

For Use With (Application) Testing and study of freshwater systems such as ponds, lakes, wetlands, rivers, streams
Grade 5+
Includes Six individual cases that house components needed for various chemical tests, sampling bottles, analysis report forms, 50 tests per factor, except nitrate (40) and pH (100), and 3 handbooks available as a download: Study of Water Quality, Limnology, and Investigating Water Problems.
Type Fresh Water Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.