Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Freshwater Mussels
SDPSCI_ED

Catalog No. S37688 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Length (English):
3 to 4 in.
4 to 5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S37688 4 to 5 in.
S37689 3 to 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S37688 Supplier Carolina™ Supplier No. S224844

For use in exploring bivalve anatomy. Carolina™ Freshwater Mussels belongs to Unio or Anodonta sp.

  • Freshwater mollusks
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Length (English) 4 to 5 in.
Length (Metric) 10.1 to 12.7 cm
Quantity 10/Pail
Packaging Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.