Learn More
STEMCELL Technologies FreSR™-S, Size: 50 mL
SDP
Catalog No. NC1250872
Encompass
FreSR™-S is a defined, serum-free, and animal component-free medium for the cryopreservation of human pluripotent stem cells (hPSCs) as single cells. This complete and ready-to-use medium is recommended for hPSCs cultured in mTeSR™1, mTeSR™ Plus, TeSR™2, or TeSR™-E8™. Cryopreserved hPSCs should be stored at -135°C (liquid nitrogen) or colder.
Catalog No. NC1250872 Supplier STEMCELL Technologies Supplier No. 05859
FreSR™-S is a defined, serum-free, and animal component-free medium for the cryopreservation of human pluripotent stem cells (hPSCs) as single cells. This complete and ready-to-use medium is recommended for hPSCs cultured in mTeSR™1, mTeSR™ Plus, TeSR™2, or TeSR™-E8™. Cryopreserved hPSCs should be stored at -135°C (liquid nitrogen) or colder.