Learn More
CPC Scientific DABCYL-Glu-Arg-Nle-Phe-Leu-Ser-Phe-Pro-EDANS (trifluoroacetate salt) 5MG
SDP
Catalog No. 502109694
Encompass
Useful FRET substrate for a continuous fluorescence-based assay of the malaria aspartyl protease (plasmepsin). The peptide sequence ERNleFLSFP is derived from the cleavage site present in hemoglobin, with Nle as a substitution for Met to avoid potential oxidation related problems.SEQUENCE: DABCYL-Glu-Arg-Nle-Phe-Leu-Ser-Phe-Pro-EDANS (trifluoroacetate salt)ONE-LETTER SEQUENCE: ERNleFLSFPMOLECULAR FORMULA: C76H98N16O15S1MOLECULAR WEIGHT: 1507.78STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [263718-22-5]SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:M.Pennington et al., Innovation and Perspectives in Solid Phase Synthesis & Combinatorial Libraries, 5th International Symposium, London, p. 367, R.Epton, ed., Mayflower Scientific, (1998)S.Jiang et al., Antimicrob. Agents Chemother., 45, 2577 (2001)M.Stadler et al., J. Antibiot., 58, 775 (2005)SKU(s): MALA-001A, MALA-001B
Catalog No. 50-210-9694 Supplier CPC Scientific Supplier No. MALA001B
Useful FRET substrate for a continuous fluorescence-based assay of the malaria aspartyl protease (plasmepsin). The peptide sequence ERNleFLSFP is derived from the cleavage site present in hemoglobin, with Nle as a substitution for Met to avoid potential oxidation related problems.SEQUENCE: DABCYL-Glu-Arg-Nle-Phe-Leu-Ser-Phe-Pro-EDANS (trifluoroacetate salt)ONE-LETTER SEQUENCE: ERNleFLSFPMOLECULAR FORMULA: C76H98N16O15S1MOLECULAR WEIGHT: 1507.78STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [263718-22-5]SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:M.Pennington et al., Innovation and Perspectives in Solid Phase Synthesis & Combinatorial Libraries, 5th International Symposium, London, p. 367, R.Epton, ed., Mayflower Scientific, (1998)S.Jiang et al., Antimicrob. Agents Chemother., 45, 2577 (2001)M.Stadler et al., J. Antibiot., 58, 775 (2005)SKU(s): MALA-001A, MALA-001B