Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FRETS-VWF73 | 97.3% | 8314.28 | 1 MG
SDP

Catalog No. 5000399618
Encompass_Preferred

FRETS-VWF73 is a 73-amino-acid peptide that serves as a fluorogenic substrate for the ADAMTS13 assay (Ex = 340 nm; Em = 450 nm). It is utilized as a predictive tool for thrombotic thrombocytopenic purpura and is intended for research use only.

  • Functions as a fluorogenic substrate for ADAMTS13 assay
  • Predictive tool for thrombotic thrombocytopenic purpura
  • Excitation wavelength: 340 nm
  • Emission wavelength: 450 nm
  • Purity: 97.3%
  • Molecular weight: 8314.28
  • Appearance: Solid, light yellow to yellow
  • Solubility: 25 mg/mL in DMSO

Catalog No. 50-003-99618 Supplier Medchemexpress LLC Supplier No. HYP5354A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FRETS-VWF73 is a 73-amino-acid peptide that serves as a fluorogenic substrate for the ADAMTS13 assay (Ex = 340 nm; Em = 450 nm). It is utilized as a predictive tool for thrombotic thrombocytopenic purpura and is intended for research use only.

  • Functions as a fluorogenic substrate for ADAMTS13 assay
  • Predictive tool for thrombotic thrombocytopenic purpura
  • Excitation wavelength: 340 nm
  • Emission wavelength: 450 nm
  • Purity: 97.3%
  • Molecular weight: 8314.28
  • Appearance: Solid, light yellow to yellow
  • Solubility: 25 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.