Learn More
Medchemexpress LLC FRETS-VWF73 TFA | 97.4% | 9340.61 | 5 MG
SDP
Catalog No. 5000399678
Encompass_Preferred
FRETS-VWF73 TFA is a 73-amino-acid peptide that functions as a fluorogenic substrate for ADAMTS13 assay.
  • 73-amino-acid peptide
  • Fluorogenic substrate for ADAMTS13 assay
  • Excitation at 340 nm
  • Emission at 450 nm
Catalog No. 50-003-99678 Supplier Medchemexpress LLC Supplier No. HYP5354A5MG
FRETS-VWF73 TFA is a 73-amino-acid peptide that functions as a fluorogenic substrate for ADAMTS13 assay.
  • 73-amino-acid peptide
  • Fluorogenic substrate for ADAMTS13 assay
  • Excitation at 340 nm
  • Emission at 450 nm