Freudenberg™ Buckets and Downpress Wringer Combination

Manufacturer: Freudenberg Household Prod LP 131582

 View more versions of this product

Catalog No. 19-200-630

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Capacity (English) 6.5 to 8 gal.
    Capacity (Metric) 24.6 to 30.3L
    Capacity 6.5 to 8 gal., 24.6 to 30.3L
    Color Yellow
    For Use With (Application) Designed to meet the demanding requirements of the most sanitary environments
    For Use With (Equipment) Fits all mop sizes
    Includes Gears and pressure plates with Fiber reinforced nylon, single bail carry handle, durable ball bearing casters, downward pressure wringer
    Material Plastic
    Product Type Bucket and Wringer
    Quantity 1 Case

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit