Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ CE Trolley Pre-Mop Box

Catalog No. 19200675 Shop All Vileda Professional Products
Catalog No. 19-200-675 Supplier Vileda Professional™ Supplier No. 120774
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Color coding clips.

For use with CE Bucket Trolley or CE Pre-Prepared Trolley

  • Autoclavable
  • Stacked
  • Does not include lid and sieve
  • Gray
  • Size: 15 x 11 x 8 in. (39 x 28 x 20 cm)

Specifications

Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.