Learn More
Vileda Professional™ CE Flat Sponge Mop Refill with Polyester Lamination, Non-sterile
Catalog No. 19032164 Shop All Vileda Professional Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
19-032-164 Autoclavable
1 options
Catalog No. 19-032-164 Supplier Vileda Professional™ Supplier No. 132123
Add to Cart
Add to Cart
Specifically for controlled environment industry allowing for 25% liquid absorption and two times more liquid release. Vileda professional™ CE Flat Sponge Mop Refill with Polyester Lamination, Non-sterile fits common plastic and stainless steel flat mopping frames for effective cleaning.
  • Features polyester lamination for tear-free application of disinfectants, cleaning and spill absorption
  • Laminated to foam without the use of adhesives to reduce contamination
  • Fits on UltraSpeed frame (40cm)
  • Suitable for autoclave, gamma and ETO sterilization
Specifications
Product Type CE Flat Sponge Mop Refill
Sterility Non-sterile
Autoclavable Autoclavable
Size 40 cm
Quantity 50/Cs.
Material Sponge with Polyester Lamination