Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ CleanTech™ Duo Microfiber Laminated Mop, Non-sterile

Catalog No. 19200661 Shop All Vileda Professional Products
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19-200-661 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-661 Supplier Vileda Professional™ Supplier No. 141886
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For cleaning/disinfecting floors, walls and ceilings. Vileda Professional CleanTech Duo Microfiber Laminated Mop, Non-sterile are designed for use in cleanrooms up to class ISO 5/GMP A and B/FED209D Class 100.

  • Microfiber laminated cleanroom foam
  • Double sided for effective cleaning and disinfecting
  • Adhesive-free lamination techniques used to prevent additional contamination to products
  • Suitable for bucket and pre-prepared method
  • Proved bacteria reduction of 99.99%

Specifications

Product Type Cleanroom Sponge Mop
Color Blue
Sterility Non-sterile
Size 50 cm
For Use With (Equipment) Telescopic Handle, Swep Duo Plus Mop Frame, Color-coded Telescopic Handles, Evolution Telescopic Handles
For Use With (Application) Floors, Walls and Ceilings, ISO 5, LAR, Production, Labs
Material Microfiber Laminated Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.