Learn More
Vileda Professional™ ClickSpeed™ Durable Mop Refill
Catalog No. 19200696 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
19-200-696 Gray
1 options
This item is not returnable. View return policy
Catalog No. 19-200-696 Supplier Vileda Professional™ Supplier No. 138695
This item is not returnable. View return policy
ClickSpeed™ is the new, easy way of mopping! When the cleaning environment and conditions consist mainly of light to medium soiled floors, ClickSpeed™ is the ideal choice. All you need is the unique magnetic frame, a Vileda Professional™ telescopic handle plus a durable or disposable mop, then you are up and ready to clean!
  • Easily assembled
  • No pockets or Velcro™ to trap dirt and bacteria
  • in.No Hands in. mop attaching and removing
  • Easy pre-preperation of mops
  • Microfiber allows for excellent cleaning results
  • Durable mop is launderable up to 300 times
  • Lower water consumption and chemicals needed
Specifications
Dimensions 44 x 20 cm
Product Type ClickSpeed Microfiber Mop
Color Gray