Description
- Made with Recycled PET bottles
- Microfiber technology minimizes linting
- Strength and durability result in long life span
- Constructed using four ply yarn- highly resistant to chemicals
- Looped-end construction alllows for maximum floor coverage while eliminating tangling, fraying and linting
Specifications
Specifications
| Product Type | Cleanroom Mop |
| Color | Green |
| Resistant To | Chemicals |
| Material | PET Synthetic Fiber |