Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ MicroTech™ Pocket Mop

Catalog No. 19200681 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-200-681 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-681 Supplier Vileda Professional™ Supplier No. 144268

missing translation for 'validMainframeMsg'

Hygiene and performance without compromise! The Vileda Professional™ pocket mop system is ideal for bucket or pre-prepared methods. The lightweight handle and frame used within the pocket mop system provides easy and effective cleaning and improved ergonomics. Different mops provide a solution for every application.

  • Low friction mop which is ideal for use on unstructured floors, tiles, etc. as the yarn construction takes the shape of any floor removing trapped dirt
  • Eliminate the need for dust mopping while dusting/wet mopping in one step
  • For use with dipping/flat sieve systems and pre-prepared mopping applications
  • Fits on CombiSpeed™ frame in addition to other common industry frames

Specifications

Dimensions 40 cm
Product Type MicroTech Pocket Mop
Color Gray
For Use With (Equipment) For use with Dipping/Flat Sieve Systems and Pre-prepared Mopping Applications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.