Learn More
Vileda Professional™ SanTech™ Duo Polyester Laminated Mop, Non-sterile
Catalog No. 19200660 Shop All Vileda Professional Products
Change view
Click to view available options
Material:
Polyester Laminated Foam
Catalog No. Material
19-200-660 Polyester Laminated Foam
1 options
Catalog No. 19-200-660 Supplier Vileda Professional™ Supplier No. 141887
For disinfecting floors, walls and ceilings. Vileda Professional SanTech Duo Polyester Laminated Mop, Non-sterile are designed for use in cleanrooms up to class ISO 5/GMP A & B/FED209D Class 100.
  • Made of polyester laminated cleanroom foam
  • Adhesive-free lamination techniques used to prevent additional contamination to products
  • Suitable for bucket and pre-prepared method
  • Better liquid absorbency
Specifications
Product Type Cleanroom Sponge Mop
Sterility Non-sterile
Size 50 cm (19.7 in.)
For Use With (Equipment) Telescopic Handle, Swep Duo Plus Mop Frame, Color-coded Telescopic Handles, Evolution Telescopic Handles
For Use With (Application) Floors, Walls and Ceilings, ISO 5, LAR, Production, Labs
Material Polyester Laminated Foam