Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ CE Dip-N-Go Sieve

Catalog No. 19200671 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-200-671 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-200-671 Supplier Vileda Professional™ Supplier No. 141928
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dip-N-Go Sieve for flat mops. Can be used on either CE Bucket Trolley or CE Pre-Prepared Trolley

  • For use when high disinfectant times are needed or deep wringing is not required
  • Can be used with either CE Pre-Prepared trolley or CE Bucket Systems Trolley
  • Stainless Steel
  • Autoclavable

Specifications

Product Type CE Dip-N-Go Sieve
Material Stainless Steel
Autoclavable Autoclavable
Color Gray
Dimensions 21 x 7 x 2 in. (54 x 17 x 5 cm)
For Use With (Equipment) CE Bucket Trolley
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.