Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ Winger™ Looped End Wet Mop

Catalog No. 19200617 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-200-617 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-617 Supplier Vileda Professional™ Supplier No. 131613
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vileda Professional™ understands that using the right mop for a specific application is very important. We have provided you with an offering that will meet your needs, whatever they may be!

  • Ideal for all production facilities
  • Superior strength and absorbency- no cotton used
  • Looped-end construction allows for maximum floor coverage while eliminating tangling, fraying and linting

Specifications

Product Type Mop
Color Blue
For Use With (Application) For all production facilities, use blue mops in coordination with white mops to avoid cross contamination issues
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.