Learn More
Labrepco Inc Upright Freezer Drawer Rack, 15 Box Capacity, Corrosion-Resistant Stainless Steel, 17 lb
SDP
Catalog No. NC3304670
Encompass_Preferred
The Upright Freezer Drawer Rack is designed for organized storage of biological samples at ultra-low temperatures, accommodating 15 polycarbonate boxes with 81 cell dividers each. Its convenient drawer-style access minimizes cold air loss, ensuring sample integrity. The construction from corrosion-resistant stainless steel adds durability vital for demanding cold storage environments. Ideal for laboratories, it combines efficiency and systematic organization for rapid identification of cryovials.
Catalog No. NC3304670 Supplier Labrepco Inc Supplier No. UDF352P81
The Upright Freezer Drawer Rack is designed for organized storage of biological samples at ultra-low temperatures, accommodating 15 polycarbonate boxes with 81 cell dividers each. Its convenient drawer-style access minimizes cold air loss, ensuring sample integrity. The construction from corrosion-resistant stainless steel adds durability vital for demanding cold storage environments. Ideal for laboratories, it combines efficiency and systematic organization for rapid identification of cryovials.