Learn More
Aviva Systems Biology Corp FRG1 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP54730_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Frg1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-962