Learn More
Vernier™ Friction Pad
SDPSCI_ED
Catalog No. S16233ND Shop All Vernier Software Products
Change view
Click to view available options
Description: Low friction
Catalog No.
S16233ND Description: Low friction
1 options
Catalog No. S16233ND Supplier Vernier™ Supplier No. PADVDS
Add to Cart
Add to Cart
Includes
Friction Pad
After working hard to make our dynamics carts very low friction, we've now added some friction back, so that you and your students can study friction in a controlled way. Not compatible with newer plastic dynamics carts.
  • Low friciton
  • Students can study friction in a controlled way.
  • Add a Cart Friction Pad to a Dynamics System, and watch the carts move under very consistent friction force.
  • Setting the friction amount is as simple as turning a knob.
Warranty and Services
5-year limited warranty
Specifications
Description Low friction
For Use With (Application) Educational use only
For Use With (Equipment) Dynamics System (2005 to 2015 models), Motion Encoder System, Plunger Cart, Standard Cart