Learn More
Grainger Approved Vendor Friction-Ring Stem Caster, Swivel, Polyolefin, 2 in. Wheel, 1 ea
Catalog No. 502792522
Encompass_Preferred
This swivel friction-ring stem caster is designed for use with office furniture, including chairs and file cabinets. It features a durable construction with a steel frame and a polyolefin wheel, providing reliable mobility for various office and light-duty industrial applications.
  • Swivel design with single ball raceway for smooth movement
  • Non-marking black wheel material
  • Friction-ring stem type
  • Operating temperature range of -20°F to +130°F
  • 75 lbs. load capacity
  • Includes wheel hood
Catalog No. 50-279-2522 Supplier Grainger Supplier No. 19YX27
This swivel friction-ring stem caster is designed for use with office furniture, including chairs and file cabinets. It features a durable construction with a steel frame and a polyolefin wheel, providing reliable mobility for various office and light-duty industrial applications.
  • Swivel design with single ball raceway for smooth movement
  • Non-marking black wheel material
  • Friction-ring stem type
  • Operating temperature range of -20°F to +130°F
  • 75 lbs. load capacity
  • Includes wheel hood