Learn More
AdipoGen Fridamycin B
SDP
Supplier:  AdipoGen BVT0470M001
Encompass_Preferred
Chemical. Formula C25H26O10. MW 486.5. Isolated from Streptomyces parvulus. C-glycosidic angucyclinone derivative. Antibiotic. Antitumor agent. Antibacterial agent. Isomer of fridamycin A Prod. No. BVT-0469.
Catalog No. 50-256-1888