Learn More
AdipoGen Fridamycin E
SDP
Supplier:  AdipoGen BVT0472M001
Encompass_Preferred
Chemical. CAS 116120-54-8. Formula C19H16O7. MW 356.3. Isolated from Streptomyces parvulus. Anthraquinone derivative derived from an angucyclinone precursor. Antibiotic. Antibacterial agent.
Catalog No. 50-256-1886