Learn More
Stoelting™ Friedman Bone Rongeurs
SDP
Catalog No. 10001970 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Rongeurs; Curved; Delicate
Catalog No. Model
10-001-970 Rongeurs; Curved; Delicate
1 options
Catalog No. 10-001-970 Supplier Stoelting™ Supplier No. 52160P
Add to Cart
Add to Cart
Stoelting™ Friedman Bone Rongeurs are sharp-edged with a scoop-shaped tip used for gouging out bone.
Used to open a window in bone, often in the skull, in neurosurgery and orthopedic surgery to expose areas for operation or to aid in removal of the brain.
Specifications
For Use With (Application) Neuroscience
Length (English) 5.5 in.
Model Rongeurs; Curved; Delicate