Learn More
Fine Science Tools Friedman Rongeur, 14cm
SDP
Catalog No. NC9690416
Encompass
Tip Shape: Curved/ Cup Size: 2.5mm/ Alloy / Material: Stainless Steel/ Length: 13cm/ Joint Action: Single. Strong and large jaws are ideal for use on the harder, outer regions of the spinal cord as well as for gaining access to delicate structures encased in bone.
Catalog No. NC9690416 Supplier Fine Science Tools Supplier No. 1600014
Tip Shape: Curved/ Cup Size: 2.5mm/ Alloy / Material: Stainless Steel/ Length: 13cm/ Joint Action: Single. Strong and large jaws are ideal for use on the harder, outer regions of the spinal cord as well as for gaining access to delicate structures encased in bone.