Learn More
Electron Microscopy Sciences Frigimat Dry Ice Maker (Us Model)
SDP
Catalog No. 5033528
Encompass_Preferred
The Frigimat Dry Ice Maker eliminates the need to buy large quantities of dry ice when only a few blocks are needed. It employs the Joule-Thomson effect to produce a solid 454 gm (16 oz.) block of dry ice in one to two minutes. A brass relief valve safely vents excess gas. Removing the dry ice block requires just 30 seconds. Each 50 lb. cylinder of liquid CO2 will make 8 to 9 blocks of dry ice, each measuring 8.5cm (3 1/8") x 11cm (4 3/8").
The unit is made from epoxy-coated steel with an aluminum ice chamber and brass valves with four non-skid rubber feet. Shipping weight: 5.5kg (12 lbs) and includes a 1.8m (6 ft) nickel-plated copper connecting tube with standard GCA 320 (0.830" – 14) thread couplings for easy attachment to an LCO2 cylinder.
Catalog No. 50-335-28 Supplier Electron Microscopy Sciences Supplier No. 6205610
The Frigimat Dry Ice Maker eliminates the need to buy large quantities of dry ice when only a few blocks are needed. It employs the Joule-Thomson effect to produce a solid 454 gm (16 oz.) block of dry ice in one to two minutes. A brass relief valve safely vents excess gas. Removing the dry ice block requires just 30 seconds. Each 50 lb. cylinder of liquid CO2 will make 8 to 9 blocks of dry ice, each measuring 8.5cm (3 1/8") x 11cm (4 3/8").
The unit is made from epoxy-coated steel with an aluminum ice chamber and brass valves with four non-skid rubber feet. Shipping weight: 5.5kg (12 lbs) and includes a 1.8m (6 ft) nickel-plated copper connecting tube with standard GCA 320 (0.830" – 14) thread couplings for easy attachment to an LCO2 cylinder.