Learn More
Teknova Friis Agar Plates. 60mm Plates, 20 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387704
Encompass
Yeast Extract 0.5570%,DEAE Dextran 0.0103%,Brain Heart Infusion Broth 0.1850%,PPLO Broth Base 0.4100%,Sodium Phosphate Dibasic Anhydrous 0.0041%,Potassium Phosphate Monobasic Anhydrous 0.0032%,Calcium Chloride Anhydrous 0.0096%,Sodium Chloride 0.4300%,Potassium Chloride 0.0215%,Magnesium Chloride Anhydrous 0.0054%,Magnesium Sulfate Anhydrous 0.0054%,Noble Agar 1.8000%
Catalog No. 50-238-7704 Supplier Teknova Supplier No. F19004PK
Yeast Extract 0.5570%,DEAE Dextran 0.0103%,Brain Heart Infusion Broth 0.1850%,PPLO Broth Base 0.4100%,Sodium Phosphate Dibasic Anhydrous 0.0041%,Potassium Phosphate Monobasic Anhydrous 0.0032%,Calcium Chloride Anhydrous 0.0096%,Sodium Chloride 0.4300%,Potassium Chloride 0.0215%,Magnesium Chloride Anhydrous 0.0054%,Magnesium Sulfate Anhydrous 0.0054%,Noble Agar 1.8000%