Learn More
Teknova Friis Media Broth. 1000mL, Sterile
SDP
Catalog No. NC1702014
Encompass
0.522% Mycoplasma Broth Base,0.492% Brain Heart Infusion,0.600% Yeast Extract,0.480% Sodium Chloride,0.024% Potassium Chloride,0.009% Disodium Phosphate,0.004% Calcium Chloride,0.004% Monopotassium Phosphate,0.003% Magnesium Sulfate,0.001% Phenol Red
This item is not returnable. View return policy
Catalog No. NC1702014 Supplier Teknova Supplier No. F0480
0.522% Mycoplasma Broth Base,0.492% Brain Heart Infusion,0.600% Yeast Extract,0.480% Sodium Chloride,0.024% Potassium Chloride,0.009% Disodium Phosphate,0.004% Calcium Chloride,0.004% Monopotassium Phosphate,0.003% Magnesium Sulfate,0.001% Phenol Red