Learn More
Teknova Friis Mycoplasma Base Medium. 500 grams
SDP
Supplier:  Teknova F0485
Encompass
0.6% Yeast extract, 0.52% PPLO Broth Base, 0.49% Brain Heart Infusion, 0.48% Sodium chloride, 0.024% Potassium chloride, 0.0009% Disodium phosphate, 0.004% Calcium chloride, 0.004% Monopotassium phosphate, 0.003% Magnesium sulfate, 0.001% Phenol red
Catalog No. 50-238-7331