Learn More
Sigma Aldrich Fine Chemicals Biosciences fRIO-TempR digital thermom1EA
Catalog No. 502900733
Encompass
Accuracy/-0.5 C from -20 to 70 C/-1 C from -50 to -20 C and 70 to 150 Cotherwise /-2 COn/Off switchData holdMax/Min memoryCE CertifiedBattery included
Catalog No. 50-290-0733 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5642221EA
Accuracy/-0.5 C from -20 to 70 C/-1 C from -50 to -20 C and 70 to 150 Cotherwise /-2 COn/Off switchData holdMax/Min memoryCE CertifiedBattery included