Learn More
Electron Microscopy Sciences Frisket Knife
SDP
Catalog No. 5026074
Encompass_Preferred
Designed for easy handling. The frisket Knife is lightweight and disposable. It comes with a fixed #11 scalpel blade, and a locking safety cap when it is not in use.
Catalog No. 50-260-74 Supplier Electron Microscopy Sciences Supplier No. 7761803
Designed for easy handling. The frisket Knife is lightweight and disposable. It comes with a fixed #11 scalpel blade, and a locking safety cap when it is not in use.