Learn More
Trajan Scientific VICI Valco Replacement Frits, Stainless Steel, 1/2" Frits, 2u Pore Size, 1.0mm Frit Thickness, 2 Pack
Catalog No. 501090288
Encompass
Replacement Frits, Stainless Steel, 1/2" Frits, 2u Pore Size, 1.0mm Frit Thickness, 2 Pack
Catalog No. 50-109-0288 Supplier Trajan Scientific Supplier No. 2FR82
Replacement Frits, Stainless Steel, 1/2" Frits, 2u Pore Size, 1.0mm Frit Thickness, 2 Pack