Learn More
IDEX Health & Science Upchurch FRIT-IN-A-FERRULE 10UM SUPE
Catalog No. NC1762171
Encompass_Preferred
Frit-in-a-Ferrule 10um Super Flangeless Tefzel (ETFE)/SST Ring
Catalog No. NC1762171 Supplier IDEX Health & Science Upchurch Supplier No. P276
Frit-in-a-Ferrule 10um Super Flangeless Tefzel (ETFE)/SST Ring