Learn More
Microsolv Technology Corporation Frit, PEEK, 5um, for use with the 7.5mm All-PEEK Column Packing System. 1 EA.
Catalog No. 502271574
Encompass_Preferred
These All PEEK Frits are made for use with the All-PEEK, 7.5mm ID, HPLC Column Hardware. 0.375" (4.78mm) Diameter and 0.312" (7.9mm) Inner Diameter.
This item is not returnable. View return policy
Catalog No. 50-227-1574 Supplier Microsolv Technology Corporation Supplier No. 49333PF
These All PEEK Frits are made for use with the All-PEEK, 7.5mm ID, HPLC Column Hardware. 0.375" (4.78mm) Diameter and 0.312" (7.9mm) Inner Diameter.