Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 0.250in Ring OD, 0.7L Volume

Catalog No. 502397981 Shop All Phenomenex Products
Encompass_Preferred

Frit, S.S., 2.0m x 0.062in Dia. x 0.062in Thick, A-101X

Catalog No. 50-239-7981 Supplier Phenomenex Inc Supplier No. A101X
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Frit, S.S., 2.0m x 0.062in Dia. x 0.062in Thick, A-101X

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.