Learn More
MilliporeSigma™ Supelco™ Fritless Purge Sampler Glassware
Catalog No. 111010424 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Non-Fritted
Volume (Metric):
5 mL
Catalog No. Description Volume (Metric)
11-101-0424 Non-Fritted 5 mL
1 options
Catalog No. 11-101-0424 Supplier MilliporeSigma™ Supelco™ Supplier No. 22780
Add to Cart
Add to Cart
For use with Tekmar™ 2000/3000 & Velocity XPT Concentrators
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Tekmar™ 2000/3000, and Velocity XPT Concentrators
Quantity 1 Ea.
Volume (Metric) 5 mL
Description Non-Fritted